Documenti fondamentali
57356-U
Supelco
SPME Inlet Guide
greener alternative
About This Item
Codice UNSPSC:
41115854
NACRES:
NB.21
Materiali
aluminum
Caratteristiche più verdi
Waste Prevention
Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
tecniche
solid phase microextraction (SPME): suitable
applicazioni
food and beverages
Categoria alternativa più verde
Descrizione generale
We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Greener Chemistry. This is a Solid Phase Microextraction (SPME) technology product. SPME is one of the greener technologies for sample preparation as it is solvent free extraction technology and SPME fibres are re-usable and generates as little waste as possible, thus aligns with "Waste Prevention”, and ″Safer Solvents and Auxiliaries″. Click here for more information.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)