Documenti fondamentali
21910
SGE Syringes for Agilent/HP 7673, 7683 and 6850 ALS Autosampler, Fixed Needle
volume 5 μL, SK-5F-HP-0.47, needle size 26 ga, needle L 42 mm, pkg of 6 ea
About This Item
Codice UNSPSC:
41122001
Materiali
cone tip stainless steel needle (point style 1)
Misura ago
26 ga
Caratteristiche
needle type F (fixed)
Confezionamento
pkg of 6 ea
Produttore/marchio commerciale
SGE 001804
Lungh. ago
42 mm
Volume
5 μL
Compatibilità
for use with Agilent/HP 7673 and 7683 autosam pler
Descrizione generale
SGE syringes are designed and tested to meet critical autosampler specifications. These syringes are designed with a unique plunger to eliminate bending and seizing. The needle point style withstands multiple and fast septum injections, making it one of the most troublefree autosampler syringes available.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)