Documenti fondamentali
26889-U
SGE Syringes for Agilent/HP 7673, 7683 and 6850 ALS Autosampler, Fixed Needle
volume 10 μL, 10F-HP-0.63/0.47, needle size 23-26 ga, needle L 42 mm
About This Item
Codice UNSPSC:
41122001
NACRES:
NB.03
Materiali
cone tip stainless steel needle
Misura ago
23-26 ga
Caratteristiche
needle type F (cemented needle)
point style #1
Confezionamento
pkg of 6 ea
Produttore/marchio commerciale
SGE 002822
Lungh. ago
42 mm
Volume
10 μL
Descrizione generale
SGE syringes are designed and tested to meet critical autosampler specifications. These syringes are designed with a unique plunger to eliminate bending and seizing. The needle point style withstands multiple and fast septum injections, making it one of the most troublefree autosampler syringes available.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)