Documenti fondamentali
Z720933
Nunc® Cell Factory accessories
Start-up Kit, sterile, 1/cs
Sinonimo/i:
cell harvesting system, closed cell culture system
About This Item
Codice UNSPSC:
41161502
NACRES:
NB.22
Sterilità
sterile
Caratteristiche
Starter kit includes: PC Connector, Airvent Filter, White Filter Adaptor Cap, Cover Caps, Tube Clamp and Silicone Tubing
Confezionamento
case of 1
pack of 1
Produttore/marchio commerciale
Nunc 170769
Descrizione generale
Starter kit includes: PC Connector, Airvent Filter, White Filter Adaptor Cap, Cover Caps, Tube Clamp and Silicone Tubing
Altre note
Cell Factory accessories for filling and emptying the cell factories with media
Note legali
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)