Documenti fondamentali
Z720917
Nunc® Cell Factory accessories
funnel, sterile, 6/cs
Sinonimo/i:
sterile funnel
About This Item
Codice UNSPSC:
41121809
NACRES:
NB.22
Sterilità
sterile
Confezionamento
pack of 1
case of 6
Produttore/marchio commerciale
Nunc 140050
Descrizione generale
  • Open interventions are reduced.
  • Ideal for aseptic methods, like fluid handing relating to filling, inoculation, feeding and harvesting of the Cell Factory system.
  • A media bag or tubing set can be plugged into any cell factory.
  • Time-saving and cost-effective.
  • Minimal cleaning processes and sterility validations.
  • γ irradiated accessories.
  • Wrapped individually.
  • Eliminates buildup of backpressure while filling the application.
  • Additional air venting options for filling and harvesting by gravity or with a peristaltic pump.
Altre note
Cell Factory accessories for filling and emptying the cell factories with media
Note legali
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)