TMO140080
Nunc® EasyFill Cell Factory filter assembly cap
high-density polyethylene, pore size 0.22 μm
Sinonimo/i:
closed cell system cap, closed cell system filter cap
About This Item
Codice UNSPSC:
41161502
NACRES:
NB.13
Prodotti consigliati
Materiali
high-density polyethylene
Confezionamento
pack of 2 ea
Produttore/marchio commerciale
Nunc 140080
Dimensione pori
0.22 μm pore size
Categorie correlate
Descrizione generale
Further the development of applications that require a more closed cell culture system using Thermo Scientific Nunc Cell Factory Accessories. Reduce the need for open interventions and support the use of aseptic methods for fluid handing relating to filling, inoculation, feeding and harvesting of the Cell Factory system.
Note legali
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
TMO140080 | 4061841580386 |