GBL103260
Grace Bio-Labs CultureWell™ adapter tray
holds 4 x 16 well chamber slides
Sinonimo/i:
chambered coverglass adapter
About This Item
Codice UNSPSC:
41122600
NACRES:
NB.22
Prodotti consigliati
Materiali
black polystyrene
Sterilità
non-sterile
Confezionamento
pack of 2 ea
Produttore/marchio commerciale
Grace Bio-Labs 103260
Lungh. × largh. esterne
127.5 mm × 85.5 mm
Descrizione generale
The adapter tray fits most universal microscope adapters and is compatible with high content screening instruments.
CultureWell Microplate Adapter Tray holds up to 4 CultureWell 16 Chambers for higher throughput.
CultureWell Microplate Adapter Tray holds up to 4 CultureWell 16 Chambers for higher throughput.
Note legali
Culturewell is a trademark of Grace Bio-Labs, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
I clienti hanno visto anche
Global Trade Item Number
SKU | GTIN |
---|---|
GBL103260-2EA | 4061832398051 |