Key Documents
EP5424702007
Eppendorf® Rotor FA-45-24-11
incl. lid, aerosol tight
About This Item
Codice UNSPSC:
41103909
NACRES:
NB.82
Caratteristiche
to hold 24 x 1.5/2.0 Ml tubes
incl. lid, aerosol tight
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Eppendorf® 5424702007
Parametri
15,000 max. RCF (21,130 xg)
tecniche
centrifugation: suitable
Compatibilità
for use with centrifuge models 5424/5424R
Note legali
Eppendorf is a registered trademark of Eppendorf AG