Key Documents
EP5409700006
Rotor for Eppendorf® 5427R Centrifuge
FA-45-12-17, aerosol tight, to hold, 12 x 5.0 mL Eppendorf Tube®, lid
About This Item
Codice UNSPSC:
41103909
NACRES:
NB.82
Caratteristiche
lid
to hold 12 x 5.0 mL Eppendorf Tube®
aerosol tight
fixed angle rotor
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Eppendorf® 5409700006 (Global)
Parametri
14,000 rpm
20,600 × g max. RCF
tecniche
centrifugation: suitable
Compatibilità
for use with centrifuge models 5427R
Note legali
Eppendorf is a registered trademark of Eppendorf AG