BMSIPD960015PTFB
BeadBlaster™ 96 PTFE Grinding ball with Steel core
bead diam. 15 mm, BeadBlaster™ 96
About This Item
Codice UNSPSC:
41100000
NACRES:
NA.82
Prodotti consigliati
Materiali
PTFE (with steel core)
Confezionamento
pack of 2
Produttore/marchio commerciale
Benchmark Scientific IPD9600-15PTFB
Diam. particelle
15 mm
Compatibilità
BeadBlaster™ 96
Categorie correlate
Descrizione generale
PTFE is the least hard of the grinding media materials and is chosen for its inert properties. PTFE is metal-free and non-stick. It′s high temperature resistance expands it′s use to cryogrinding. PTFE grinding media is used for applications in the pharmaceutical field as well as for plants and other biological samples. The 15mm PTFE grinding balls have steel cores for a higher energy input.
Applicazioni
Homogenization of biological and pharmaceutical samples
Caratteristiche e vantaggi
- Inert material
- Non-stick
- Wide temperature stability
Note legali
BeadBlaster is a trademark of Benchmark Scientific, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)