Z173185
Filter funnel with drip-tip and two glass stopcocks, one on side-arm
capacity 75 mL, male joint: ST/NS 14/20, female joint: ST/NS 14/20
About This Item
Codice UNSPSC:
41104900
NACRES:
NB.43
Prodotti consigliati
Materiali
borosilicate glass
Etichetta
X
Giunto
female joint: ST/NS14/20
male joint: ST/NS14/20
Produttore/marchio commerciale
Aldrich
Capacità
75 mL
Dimensione pori
25-50 μm porosity
Categorie correlate
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Articoli
Versatile system for chemical reactions, extractions, and transfers of solids and liquids in laboratories.
Global Trade Item Number
SKU | GTIN |
---|---|
Z173185-1EA | 4061837617058 |