HS120627
smartPans™ Cahn Style Aluminum Microbalance Weigh Pan
pan diam. 25 mm
About This Item
Código UNSPSC:
41121812
NACRES:
NB.35
Produtos recomendados
Materiais
aluminum
embalagem
pack of 20 ea
fabricante/nome comercial
Heathrow Scientific 120627
Altura
3.6 mm
diâmetro do tacho
25 mm
Descrição geral
The Cahn style aluminum weigh pan is ideal for microbalances, organic micro elementary and thermal analysis, and pharmaceutical use.
Características e benefícios
- Ideal for microbalances, organic micro elementary and thermal analysis, and pharmaceutical use.
- Made of Ultra High Vacuum (UVH) Foil - “A” wettable and completely free from oils normally found on other grades of aluminum foil
- Foil meets ASTM B-479, specification 3.1.4 and 10.3.1
- Cahn style” accessory pans are compatible with most commercially available analyzers and microbalances
- Pan is recyclable
Informações legais
smartPans is a trademark of LevGo Inc.
Escolha uma das versões mais recentes:
Certificados de análise (COA)