Documentos Principais
Z685267
Platinum crucible, with reinforced rim
capacity 10 mL
About This Item
Código UNSPSC:
41122200
NACRES:
NB.43
fabricante/nome comercial
Birmingham metals
capacidade
10 mL
Descrição geral
Platinum labware has excellent resistance to acids such as acetic, hydrochloric, hydrofluoric, hydroiodic, nitric, selenic, and sulfuric, and to oxidation at high temperatures. These platinum crucibles can be cleaned by boiling in a suitable solvent, for the removal of substances which have alloyed with the surface, scour with alumina powder. Having a reinforced rim means that the rim of the crucible is thicker than the walls to make it stronger; this is where the crucible can be gripped if tongs are used during the handling process.
Escolha uma das versões mais recentes:
Certificados de análise (COA)
Visite a Biblioteca de Documentos
Os clientes também visualizaram
Slide 1 of 10
1 of 10
Platinum evaporating dish capacity 20 mL
Z562130
Platinum evaporating dish
Platinum crucible standard, capacity 25 mL
Z562076
Platinum crucible
Platinum combustion boat capacity 3 mL, with handle
Z685364
Platinum combustion boat
Platinum crucible standard, capacity 50 mL
Z562114
Platinum crucible
Platinum crucible lid fits 30 mL crucible
Z562106
Platinum crucible lid
Platinum crucible lid fits 25 mL crucible
Z562084
Platinum crucible lid
Precision crucible tongs L 20 in. (508 mm)
Z225282
Precision crucible tongs
Global Trade Item Number
SKUGTIN
Z685267-1EA4061833571910