Documentos Principais
Z680265
Titanium tweezers
Style 7
About This Item
Código UNSPSC:
27112105
NACRES:
MA.01
fabricante/nome comercial
Aven
total C
4.5 in. (115 mm)
Descrição geral
Titanium tweezers do not shed particles like stainless steel tweezers. Titanium is light weight, high strength, non-magnetic material.
  • Lightweight (high strength to weight ratio)
  • Zero magnetic retentively
  • Temperature resistant to 800 °F
  • Chemical and corrosion resistant
  • Non-solder adhesive
  • Bio-compatible (maintain cell integrity, no inflammatory response)
Typical application: handling of sensitive components in cleaning/chemical processes at high temperatures, histology, biology, forensics, electron microscopy etc.
Escolha uma das versões mais recentes:
Certificados de análise (COA)
Visite a Biblioteca de Documentos
Os clientes também visualizaram
Slide 1 of 10
1 of 10
Tweezers Style #7, sharp, hooked, anti-magnetic stainless steel
T4912
Tweezers
Tweezers Style #7, sharp, hooked, PTFE-coated, anti-magnetic stainless steel
T6165
Tweezers
Tweezers Style #7, sharp, hooked, non-magnetic alloy (tips may be restraightened when bent)
T4787
Tweezers
Artis Tweezer Style 7-SA
Z742677
Artis Tweezer
Artis tweezer Style 5-SA
Z742676
Artis tweezer
Tweezers Style #00, sharp, anti-magnetic stainless steel
T5415
Tweezers
Disposable forceps dark blue, L 110 mm, straight tip
Z708356
Disposable forceps
Tweezers Style #3, very sharp, PTFE-coated, anti-magnetic stainless steel
T5540
Tweezers
Stainless steel forceps Fine tip, curved
Z168785
Stainless steel forceps
Global Trade Item Number
SKUGTIN
Z680265-1EA4061826711422