About This Item
Código UNSPSC:
27112105
NACRES:
MA.01
Produtos recomendados
fabricante/nome comercial
Aven
total C
4.75 in. (121 mm)
Categorias relacionadas
Descrição geral
Titanium tweezers do not shed particles like stainless steel tweezers. Titanium is light weight, high strength, non-magnetic material.
Typical application: handling of sensitive components in cleaning/chemical processes at high temperatures, histology, biology, forensics, electron microscopy etc.
- Lightweight (high strength to weight ratio)
- Zero magnetic retentively
- Temperature resistant to 800 °F
- Chemical and corrosion resistant
- Non-solder adhesive
- Bio-compatible (maintain cell integrity, no inflammatory response)
Typical application: handling of sensitive components in cleaning/chemical processes at high temperatures, histology, biology, forensics, electron microscopy etc.
Escolha uma das versões mais recentes:
Certificados de análise (COA)
Os clientes também visualizaram
Global Trade Item Number
SKU | GTIN |
---|---|
Z680222-1EA | 4061826711385 |