Z567078
NMR tube carrier
for tubes, 5 mm x 7 or 8 in.
About This Item
Código UNSPSC:
41100000
NACRES:
SB.32
Produtos recomendados
Características
for tubes 5 mm x 7 or 8 in.
Descrição geral
Unbreakable, NMR tube carrier provides protection for the tube and lab personnel when transporting samples. The clear, shatter-resistant polycarbonate case allows for visual sample identification and inspection prior to opening. Natural rubber plugs on both ends of the case provide impact absorption if accidentally dropped.
NMR tubes are held securely in place inside the PC case by the bottom loading rubber plug. Carrier holds one 5 mm diameter tube, 7 or 8 in. L.
NMR tubes are held securely in place inside the PC case by the bottom loading rubber plug. Carrier holds one 5 mm diameter tube, 7 or 8 in. L.
Escolha uma das versões mais recentes:
Certificados de análise (COA)
Os clientes também visualizaram
Global Trade Item Number
SKU | GTIN |
---|---|
Z567078-5EA | 4061837637049 |