Documentos Principais
Z225509
Flask brush
Black nylon
About This Item
Código UNSPSC:
47131605
NACRES:
NB.35
Descrição geral
Stiff 41/2 × 13/4in. brush mounted on a preformed, plastic handle that adapts itself to the curvature of the flask. Overall L: 16 in. Fits through 24/40 joints.
Escolha uma das versões mais recentes:
Certificados de análise (COA)
Visite a Biblioteca de Documentos
Os clientes também visualizaram
Slide 1 of 6
1 of 6
Laboratory brushes Micro tube, L 7 in., brush diam. × L 3/16- 1/2 in. × 3 in.
Z281247
Laboratory brushes
Laboratory brushes Burette, L 36 in., brush diam. × L 3/4 in. × 3 1/4 in.
Z281379
Laboratory brushes
Laboratory brushes Centrifuge, L 12 in., brush diam. × L 1 1/2 in. × 4 1/2 in.
Z281352
Laboratory brushes
Laboratory brushes Narrow tube, L 12 in., brush diam. × L 1/2 in. × 3 in.
Z281301
Laboratory brushes
Laboratory brushes Funnel, L 13 in., brush diam. × L 2 1/4 in. × 6 in.
Z281336
Laboratory brushes
Laboratory brushes Burette, L 30 in., brush diam. × L 1/2 in. × 3 1/4 in.
Z281360
Laboratory brushes
Global Trade Item Number
SKUGTIN
Z225509-1EA4061832331898