Key Documents
Z281344
Laboratory brushes
Flask, L 12 in., brush diam. × L 1 3/8 in. × 3 1/2 in.
About This Item
Código UNSPSC:
47131605
NACRES:
NB.35
embalagem
pkg of 4 ea
Comprimento
12 in.
brush diam. × L
1 3/8 in. × 3 1/2 in.
Descrição geral
Brush materials resistant to acids, aromatic and aliphatic hydrocarbons, ketones, ethyl acetate esters, trichloroethylene, and hot water. FDA and USDA approved materials.
Escolha uma das versões mais recentes:
Certificados de análise (COA)
Lot/Batch Number
Lamentamos, não temos COA para este produto disponíveis online no momento.
Visite a Biblioteca de Documentos
Os clientes também visualizaram
Slide 1 of 6
1 of 6
Laboratory brushes Test tube, L 10 in., brush diam. × L 1 3/8 in. × 4 in.
Z281263
Laboratory brushes
Laboratory brushes Test tube, L 9 in., brush diam. × L 3/4 in. × 3 1/4 in.
Z281255
Laboratory brushes
Laboratory brushes Narrow tube, L 12 in., brush diam. × L 1/2 in. × 3 in.
Z281301
Laboratory brushes
Laboratory brushes Micro tube, L 7 in., brush diam. × L 3/16- 1/2 in. × 3 in.
Z281247
Laboratory brushes
Laboratory brushes Burette, L 36 in., brush diam. × L 3/4 in. × 3 1/4 in.
Z281379
Laboratory brushes