Documentos Principais
Z214639
Microscale vacuum adapter
with drip-tip
About This Item
Código UNSPSC:
41121815
NACRES:
NB.43
fabricante/nome comercial
Ace Glass 526106
Descrição geral
#7 Ace-Thred at top with a nylon bushing and FETFE® O-ring that form a compression seal with microscale condenser tube, filter funnel, bleed tube, or thermometer (6.5 to 7.0 mm diam.). Has bottom 14/10 inner joint and side hose connection. (Shown with Hirsch funnel sold separately.)
Informações legais
FETFE is a registered trademark of Ace Glass, Inc.
Escolha uma das versões mais recentes:
Certificados de análise (COA)