Documentos Principais
Z168742
Stainless steel forceps
Dumont
About This Item
Código UNSPSC:
41122405
eCl@ss:
32035504
NACRES:
NB.35
embalagem
pack of 1 ea
Comprimento
4 3/4 in.
Descrição geral
with very sharp points, for picking up minute articles
Escolha uma das versões mais recentes:
Certificados de análise (COA)
Visite a Biblioteca de Documentos
Os clientes também visualizaram
Slide 1 of 10
1 of 10
Stainless steel forceps Reverse action
Z168769
Stainless steel forceps
Stainless steel forceps Specimen, L 4 1/2 in.
Z168726
Stainless steel forceps
Stainless steel forceps Specimen, L 5 1/2 in.
Z225592
Stainless steel forceps
Stainless steel forceps Specimen, L 6 in.
Z168734
Stainless steel forceps
Bayonet forceps stainless steel
F2394
Bayonet forceps
Dressing tissue forceps L 4 3/4 in.
F3892
Dressing tissue forceps
Stainless steel forceps Specimen, L 10 in.
Z225614
Stainless steel forceps
Dressing tissue forceps L 6 in.
F4267
Dressing tissue forceps
Global Trade Item Number
SKUGTIN
Z168742-1EA4061833298886