Documentos Principais
F3767
Sigma-Aldrich
Micro-dissecting forceps
very fine point
Sinônimo(s):
dissecting forceps, tissue forceps
About This Item
Código UNSPSC:
41122405
NACRES:
NB.35
Materiais
stainless steel
embalagem
pack of 1 ea
Comprimento
4 1/2 in.
tamanho da ponta
2 mm
Descrição geral
with serrated tip
Escolha uma das versões mais recentes:
Certificados de análise (COA)
Visite a Biblioteca de Documentos
Os clientes também visualizaram
Slide 1 of 9
1 of 9
Artis Tweezer Style 7-SA
Z742677
Artis Tweezer
Dressing tissue forceps L 4 3/4 in.
F3892
Dressing tissue forceps
Bayonet forceps stainless steel
F2394
Bayonet forceps
Artis Tweezer Style AA-SA
Z742671
Artis Tweezer
Micro-dissecting scissors size 4 in., curved, sharp point stainless steel
S3271
Micro-dissecting scissors
Stainless steel forceps Sharp-point
Z168718
Stainless steel forceps
Artis Tweezer Style OO-SA
Z742670
Artis Tweezer
Tweezers Style #3C, very sharp, non-magnetic alloy (tips may be restraightened when bent)
T4162
Tweezers
Tweezers Style #5, Fine needle-sharp, Inox alloy (not anti-magnetic)
T4662
Tweezers