Documentos Principais
Z168769
Stainless steel forceps
Reverse action
About This Item
Código UNSPSC:
41122405
eCl@ss:
32035504
NACRES:
NB.35
Descrição geral
with sharp, smooth points, normally closed, easily opens with finger pressure
Escolha uma das versões mais recentes:
Certificados de análise (COA)
Visite a Biblioteca de Documentos
Os clientes também visualizaram
Slide 1 of 10
1 of 10
Stainless steel forceps Fine tip, straight
Z168777
Stainless steel forceps
Artis tweezer Style 5-SA
Z742676
Artis tweezer
Stainless steel forceps Specimen, L 8 in.
Z225606
Stainless steel forceps
Stainless steel forceps Specimen, L 4 1/2 in.
Z168726
Stainless steel forceps
Dressing tissue forceps L 6 in.
F4267
Dressing tissue forceps
Dressing tissue forceps L 8 in.
F4392
Dressing tissue forceps
Bayonet forceps stainless steel
F2394
Bayonet forceps
Stainless steel forceps Specimen, L 10 in.
Z225614
Stainless steel forceps