Z136115
Scienceware® vernier caliper
Direct-reading type
About This Item
Código UNSPSC:
41111700
NACRES:
NB.79
Produtos recomendados
embalagem
pack of 1 ea
fabricante/nome comercial
Bel-Art H13415-0000
Descrição geral
Metric and English scales with 0.1 mm and 0.001 in. subdivisions.
Metric/Imperial. Molded in light, corrosion-resistant plastic. 6 in. or 150 mm measuring capacity; parallel jaws are 1.57 in. (40 mm) deep; reading scales create no parallax. Nonconductive, fine-adjustment thumb roller, two-direction lock, inside-outside jaws, round-rod depth gauge. Upper scale reads in mm, lower scale in inches.
Informações legais
Scienceware is a registered trademark of SP Scienceware, a division of SP
Escolha uma das versões mais recentes:
Certificados de análise (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z136115-1EA | 4061832262260 |