HPLC Analysis of Catechols and Resorcinols on Ascentis® RP-Amide
![HPLC Analysis of Catechols and Resorcinols on Ascentis® RP-Amide application for HPLC](/deepweb/assets/sigmaaldrich/product/images/227/969/d6db0621-7876-485f-9dac-d07e950ae634/800/d6db0621-7876-485f-9dac-d07e950ae634.jpg)
Materiały
kolumna analityczna
Numer produktu
Opis
Cennik
CONDITIONS
column
Ascentis RP-Amide, 15 cm x 4.6 mm I.D., 5 μm particles (565324-U)
mobile phase
[A] 20 mM phosphoric acid (pH 2.0 unadjusted): [B] acetonitrile; (75:25, A:B)
flow rate
1.5 mL/min
column temp.
30 °C
detector
UV, 270 nm
injection
25 μL
sample
as indicated in 20 mM phosphoric acid (pH 2.0)
Opis
Informacje prawne
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany