GF13576934
Iron/Silicon
powder, Fe 91%/Si 9%, mean particle size 10 micron, weight 100 g, condition gas atomized
Synonym(s):
FE166012
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
form
powder
manufacturer/tradename
Goodfellow 135-769-34
weight
100 g
mean particle size
10 μm
Looking for similar products? Visit Product Comparison Guide
General description
For updated SDS information please visit www.goodfellow.com
Legal Information
Product of Goodfellow
signalword
Warning
hcodes
Hazard Classifications
Flam. Sol. 2 - Self-heat. 2
Storage Class
4.2 - Pyrophoric and self-heating hazardous materials
wgk_germany
WGK 3
flash_point_f
69.8 °F
flash_point_c
21 °C
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service