Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CF0010

Sigma-Aldrich

Acid Togni-(ImidazolylCF2CF2)-reagent

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C12H7F4IN2O2
CAS Number:
MDL number:
UNSPSC Code:
12352106

form

solid

reaction suitability

reaction type: C-C Bond Formation

Application

Acid Togni-(Imidazolyl-CF2CF2)-reagent is an electrophilic hypervalent iodine-fluoroalkyl reagent introducing the imidazolyltetrafluoroethyl fragment. It can also engage in copper mediated tandem radical fluoroalkylation-cyclization with alkenes, creating novel tetrafluorinated imidazopyridines.

Automate your fluorination reactions with Synple Automated Synthesis Platform (SYNPLE-SC002)

Legal Information

Product of CF Plus Chemicals.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

The fluoroalkylation toolbox now includes Togni reagents, hypervalent iodine perfluoroalkylation reagents, fluoroalkyl bromides, silanes, carboxylates, and sulfonyl fluorides for late stage fluoroalkylation.

The fluoroalkylation toolbox now includes Togni reagents, hypervalent iodine perfluoroalkylation reagents, fluoroalkyl bromides, silanes, carboxylates, and sulfonyl fluorides for late stage fluoroalkylation.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service