Merck
GC Analysis of Permanent Gases on Mol Sieve 5A PLOT
GC Analysis of Permanent Gases on Mol Sieve 5A PLOT suitable for GC
Materiały
kolumna analityczna
Numer produktu
Opis
Cennik
Mol Sieve 5A PLOT Capillary GC Column
L × I.D. 30 m × 0.53 mm, average thickness 30 μm
CONDITIONS
column
Mol Sieve 5A PLOT, 30 m x 0.53 mm I.D. (25463)
oven
65 °C
inj. temp.
200 °C
detector
TCD, 230 °C
carrier gas
helium, 10.0 mL/min
injection
10.0 μ, direct using a sample valve
sample
Oxygen, methane, and carbon monoxide, each at 1.0% in nitrogen