Kluczowe dokumenty
RAD122
radiello Vertical Adapter
threaded for standard use, pk of 20
About This Item
Kod UNSPSC:
41104008
NACRES:
NB.24
opakowanie
pk of 20
producent / nazwa handlowa
Radiello Code 122
metody
passive air sampling: suitable
Zastosowanie
air monitoring
environmental
industrial hygiene
Opis ogólny
For personal sampling (breathing zone assessment) a vertical adapter is available to position the Radiello diffusive body vertically on the Radiello triangular support plate.
Informacje prawne
radiello is a trademark of Institi Clinici Scientifici Maugeri
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)
Odwiedź Bibliotekę dokumentów
J Dekeirsschieter et al.
Forensic science international, 189(1-3), 46-53 (2009-05-09)
Forensic entomology uses pig carcasses to surrogate human decomposition and to investigate the entomofaunal colonization. Insects communicate with their environment through the use of chemical mediators, which in the case of necrophagous insects, may consist in the cadaveric volatile organic
Armelle Vallat et al.
Phytochemistry, 66(13), 1540-1550 (2005-06-14)
Headspace volatiles from apple-bearing twigs were collected in the field with a Radiello sampler during three different diurnal periods over the complete fruit growing season. Analyses by thermal desorption-GC-MS identified a total of 62 compounds in changing quantities, including the
Benoît Oury et al.
Journal of occupational and environmental hygiene, 3(10), 547-557 (2006-08-16)
Diffusive sampling is particularly suited to determine time-weighted average volatile organic compound (VOC) concentration in occupational hygiene and environmental air monitoring. The purpose of this study was to measure the sampling rate variation of four different samplers in a special
Rocco Rella et al.
The Science of the total environment, 487, 255-259 (2014-05-03)
The proofing treatment of the floor slabs with a solvent based material and successive treatment with epoxy resin resulted in significant air contamination in a primary school. The contamination caused illnesses in many occupants of school. The GC/MS investigations of
Anne Pennequin-Cardinal et al.
Talanta, 65(5), 1233-1240 (2008-10-31)
Radiello((R)) diffusive samplers filled with a thermally desorbable adsorbent (graphitised charcoal Carbograph 4) have been tested for the monitoring of BTEX. The sampling rates have been estimated under various controlled atmospheres in order to evaluate the effects of two factors
Produkty
Cartridge RAD147 is a stainless steel net cylinder with 100 mesh grid opening and 4.8 mm diameter, packed with 250 ± 10 mg of Tenax-TA, particle size 20-35 mesh. Phenols are trapped by adsorption and recovered by thermal desorption, analysis is performed by capillary gas chromatography and MS detection.
radiello® Diffusive Air Sampling Application - Ammonia
radiello® Diffusive Air Sampling Application - Hydrogen Sulfide
Learn about radiello® Diffusive Sampling Aldehydes Applications & Methods.
Protokoły
radiello® Diffusive Air Sampling Application - Nitrogen and Sulfur Dioxide
The adsorbing cartridge is formed by a micropore polyethylene tube filled with silica gel coated with 4,4’-dipyridylethylene and closed, at one end, by a PTFE cap. Upon exposure, acid-catalysed ozonolysis of 4,4’-dipyridylethylene leads to 4-pyridylaldehyde.
radiello® Diffusive Air Sampling Application - Volatile Organic Compounds (VOCs) Thermally Desorbed
radiello® Diffusive Air Sampling Application - Anaesthetic Gases and Vapours
Zobacz wszystko