Key Documents
RAD121
radiello Triangular Support Plate
pk of 20
About This Item
Kod UNSPSC:
41104008
NACRES:
NB.24
opakowanie
pk of 20
producent / nazwa handlowa
Radiello Code 121
metody
passive air sampling: suitable
Zastosowanie
air monitoring
environmental
industrial hygiene
Opis ogólny
A radiello diffusive sampler consists of 3 parts: the diffusive body, the adsorbent tube, and the triangular support plate. For personal air sampling, a vertical adapter can be installed. The adsorbent cartridge is removed from its glass or plastic storage tube and is inserted into the diffusive body. This is then threaded onto the triangular support plate in order to place the sampler in its sampling position.
The triangular support plate acts as both a closure and means of suspension for the Radiello diffusive body and cartridge adsorbent during passive air sampling. Each support plate is threaded for easy diffusive body attachment. Each plate includes a clip and transparent adhesive pocket to hold the Radiello barcode label.
Informacje prawne
radiello is a trademark of Institi Clinici Scientifici Maugeri
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)
Lot/Batch Number
Przepraszamy, ale COA dla tego produktu nie jest aktualnie dostępny online.
Odwiedź Bibliotekę dokumentów
Study of the dispersion of VOCs emitted by a municipal solid waste landfill
Chiriac, R., et al.
Atmospheric Environment, 43 (11), 1926-1931 (2009)
Isabel M Morales Terrés et al.
Journal of environmental management, 91(12), 2754-2762 (2010-09-03)
This paper describes a novel methodology for evaluating the extent to which petrol stations affect their surroundings. The method is based on the fact that the ratio of the concentrations of aliphatic and aromatic hydrocarbon pollutants in the air of
J Dekeirsschieter et al.
Forensic science international, 189(1-3), 46-53 (2009-05-09)
Forensic entomology uses pig carcasses to surrogate human decomposition and to investigate the entomofaunal colonization. Insects communicate with their environment through the use of chemical mediators, which in the case of necrophagous insects, may consist in the cadaveric volatile organic
Armelle Vallat et al.
Phytochemistry, 66(13), 1540-1550 (2005-06-14)
Headspace volatiles from apple-bearing twigs were collected in the field with a Radiello sampler during three different diurnal periods over the complete fruit growing season. Analyses by thermal desorption-GC-MS identified a total of 62 compounds in changing quantities, including the
Benoît Oury et al.
Journal of occupational and environmental hygiene, 3(10), 547-557 (2006-08-16)
Diffusive sampling is particularly suited to determine time-weighted average volatile organic compound (VOC) concentration in occupational hygiene and environmental air monitoring. The purpose of this study was to measure the sampling rate variation of four different samplers in a special
Produkty
Cartridge RAD147 is a stainless steel net cylinder with 100 mesh grid opening and 4.8 mm diameter, packed with 250 ± 10 mg of Tenax-TA, particle size 20-35 mesh. Phenols are trapped by adsorption and recovered by thermal desorption, analysis is performed by capillary gas chromatography and MS detection.
radiello® Diffusive Air Sampling Application - Ammonia
radiello® Diffusive Air Sampling Application - Hydrogen Sulfide
Learn about radiello® Diffusive Sampling Aldehydes Applications & Methods.
Protokoły
radiello® Diffusive Air Sampling Application - Nitrogen and Sulfur Dioxide
The adsorbing cartridge is formed by a micropore polyethylene tube filled with silica gel coated with 4,4’-dipyridylethylene and closed, at one end, by a PTFE cap. Upon exposure, acid-catalysed ozonolysis of 4,4’-dipyridylethylene leads to 4-pyridylaldehyde.
radiello® Diffusive Air Sampling Application - Volatile Organic Compounds (VOCs) Thermally Desorbed
radiello® Diffusive Air Sampling Application - Anaesthetic Gases and Vapours
Zobacz wszystko