Key Documents
58803
Cutting Wheel for TC-10
About This Item
Kod UNSPSC:
41115700
opis
TC-10
opakowanie
pkg of 3 ea
Opis ogólny
Replacement cutting wheel for SSI Model TC-10 tubing cutter. Will not fit Model TC-20.
Przypis
Replacement cutting wheels for SSI Model TC-10 tubing cutter. Will not fit Model TC-20.
Polecane produkty
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)