Key Documents
502707
PEEK Inline Microfilter
About This Item
Kod UNSPSC:
41115700
opakowanie
pkg of 1 ea
Opis ogólny
Install between 2 pieces of capillary tubing, using Microtight tubing sleeves. PEEK housing with special PEEK fitting that encases the 0.5 μm PEEK frit. Through hole 0.006 in., swept volume 0.24 μL. Includes 2 MicroFingertight nuts, and 5 frits (1 installed).
Polecane produkty
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
część zamienna
Numer produktu
Opis
Cennik
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)