Merck
22460
Supelco
Supeltex M-4 Capillary Ferrule, General Purpose
I.D. 1.2 mm, 0.50-0.75 mm Column I.D., pkg of 10 ea
About This Item
Kod UNSPSC:
41121815
opakowanie
pkg of 10 ea
śr. wewn.
1.2 mm
kompatybilność
0.50-0.75 mm Column I.D.
Opis ogólny
Supelco has refined the design of these graphite ferrules to provide a clean, sharp profile with minimal flash.
Characteristics:
Seals at one-quarter turn past fingertight. Maximum sealing surface contact, reduced risk of column contamination at installation.
Material Composition: flexible graphite
Maximum Temperature: 450 °C
Informacje prawne
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)