Kluczowe dokumenty
TTRT96P250B-EP
Eppendorf twin.tec® Real-Time PCR Plates 96
Skirted, PCR clean, volume 250 μL, blue plate (white wells), pack of 25
About This Item
Kod UNSPSC:
41106300
Materiały:
blue plate (white wells)
polypropylene wells
rozmiar:
96 well
pojemność:
250 μL
pojemność robocza:
250 μL
sterylność:
non-sterile
Właściwości:
skirt
PCR clean
Skirted
Materiały
blue plate (white wells)
polypropylene wells
sterylność
non-sterile
Właściwości
skirt
PCR clean
Skirted
opakowanie
pack of 25
producent / nazwa handlowa
Eppendorf® 951022003 NA Only
metody
PCR: suitable
rozmiar
96 well
pojemność
250 μL
pojemność robocza
250 μL
Opis ogólny
The white wells of Eppendorf’s twin.tec real-time PCR plates increase the reflection of fluorescent light in real-time PCR (qPCR) and other fluorescence assays. The increased fluorescence from these specialized qPCR plates improves the sensitivity of your assays, while also improving reproducibility and consistency of your results.
Zastosowanie
Eppendorf® twin.tec® Real-Time PCR Plates 96 are suitable for real-time polymerase chain reaction (PCR) and melting curve analysis
Cechy i korzyści
  • Polypropylene, white wells ensure better reflection of fluorescent light
  • Highly stable to mechanical stress
  • Effective sealing by raised well rims
  • PCR plates feature skirted frames
  • Optimal heat transfers due to reduced wall thickness
  • Autoclavable at 121 °C for 20 min
  • White wells for better reflection
  • High mechanical stability
  • Raised rims for effective sealing
  • Skirted and semi-skirted frames
  • Optimal heat transfer due to reduced wall thickness
  • Autoclavable (121 °C, 20 min)
  • PCR clean and Eppendorf Forensic DNA Grade s available
Informacje prawne
Eppendorf is a registered trademark of Eppendorf AG
twin.tec is a registered trademark of Eppendorf AG
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)