Key Documents
MTCP7025
MTC Bio ASPIR-8 Resevoir for 8-Channel Pipettes
new
capacity 25 mL, non-sterile, pack of 100 ea (bulk bag)
Synonim(y):
cryo vial marker, reagent dish, reagent holder
About This Item
Kod UNSPSC:
41121522
sterylność
non-sterile
opakowanie
pack of 100 ea (bulk bag)
producent / nazwa handlowa
MTC Bio P7025
pojemność
25 mL
Opis ogólny
Aspir-8 Reservoir, 25ml, 8-channel only, non-sterile, bulk pack, 100/cs
Cechy i korzyści
  • Graduated inner walls
  • Micro-trough U-shaped bottoms
  • Pouring spouts on all 4 corners
Informacje prawne
ASPIR-8 is a trademark of MTC Bio
MTC is a trademark of MTC Bio