CLS7007510
Pyrex® VISTA™ graduated cylinder, to contain, economy
volume 10 mL
Synonim(y):
akcesoria do sprzętu laboratoryjnego
About This Item
Kod UNSPSC:
41121805
NACRES:
NB.43
Polecane produkty
producent / nazwa handlowa
Corning 70075-10
pojemność
10 mL
Powiązane kategorie
Opis ogólny
These Pyrex VISTA cylinders are calibrated "to contain" (TC). They are supplied in two parts - a strong, accurate glass graduate and a detachable, sturdy plastic base which absorbs impacts and reduces breakage. Tolerances are +/- 5% of total at any point. By removing the base, the graduate can be conveniently stacked. Bumper guards are supplied with 25 mL through 100 mL inclusive. The 10 mL has a funnel top for careful pouring.
Informacje prawne
PYREX is a registered trademark of Corning, Inc.
VISTA is a trademark of Corning, Inc.
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)
Global Trade Item Number
SKU | GTIN |
---|---|
CLS7007510-12EA | 4061831702965 |