About This Item
Kod UNSPSC:
41122413
NACRES:
NB.35
Polecane produkty
rozmiar
M
dł. × śr.
211 mm × 11 mm , arm
pojemność szczęk
91-114 mm
Opis ogólny
Corrosion resistant stainless steel. Ideal for holding chromatography columns and large cylindrical glassware. A large, flat thumbscrew/worm drive permits maximum tightening or removal in minimum time.
NOTE: An additional clamp holder is required to attach clamps to frames or other apparatus.
NOTE: An additional clamp holder is required to attach clamps to frames or other apparatus.
Informacje prawne
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)