Z635553
Swiss Boy lab jacks
load ≤90 kg (with two plates, middle position), platform L × W 400 mm × 500 mm, Model 118
About This Item
Kod UNSPSC:
41122800
NACRES:
NB.35
Polecane produkty
producent / nazwa handlowa
Grauer
Parametry
≤90 kg load (with two plates, middle position)
122-498 mm vertical limit
platforma dł. × szer.
400 mm × 500 mm
kolor
gray
Opis ogólny
Podnośnik można podnieść z ładunkiem na miejscu.
These high quality Swiss made jacks have massive cast aluminum frames with a chemically resistant anodised finish. Swiss boy jacks are designed to carry heavy loads safely. Available either in standard size or with extended carrying plates for large pots or pans. Units carrying more that 90kg are also available with a motorized lift adjustment for enhanced convenience. The max load is for the middle position. Supplied with top and bottom plates.
Products with motorized jacks are available in Europe only.
Products with motorized jacks are available in Europe only.
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z635553-1EA | 4061833581599 |