Z567272
Ace round-bottom pressure flask with thermowell
with Ace-Thred 15 PTFE front-seal plug, capacity 250 mL
About This Item
Kod UNSPSC:
41121804
NACRES:
NB.43
Polecane produkty
Materiały
borosilicate glass
round bottom flask
single-neck
sterylność
non-sterile
Właściwości
closure type PTFE plug seal
opakowanie
pack of 1
producent / nazwa handlowa
Ace Glass 8417-07
pojemność
250 mL
Średnica
82 mm
Powiązane kategorie
Opis ogólny
Heavy-wall borosilicate glass flasks are pressure rated to 60 psig at 120°C. The rounded bottom facilitates use in heating mantles and for other round-bottom flask applications. Flasks are supplied complete with #15 Ace-Thred PTFE front seal bushing and FETFE® O-ring.
Replacement FETFE O-rings:
Ace-Thred #Front SealBack Seal
7 Z504661 Z504742
15 Z504858 Z505021
25 Z505056 Z505110
CAUTION: This glassware is not guaranteed against breakage caused by pressure or vacuum. These flasks should never be used if scratched or otherwise damaged.
Replacement FETFE O-rings:
Ace-Thred #Front SealBack Seal
7 Z504661 Z504742
15 Z504858 Z505021
25 Z505056 Z505110
CAUTION: This glassware is not guaranteed against breakage caused by pressure or vacuum. These flasks should never be used if scratched or otherwise damaged.
Informacje prawne
FETFE is a registered trademark of Ace Glass, Inc.
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z567272-1EA | 4061832351131 |