Z531138
Aldrich® coiled condenser
unjacketed, L 300 mm, joint: ST/NS 14/20
About This Item
Kod UNSPSC:
41121800
NACRES:
NB.43
Polecane produkty
Połączenie
joint: ST/NS14/20
Długość
300 mm
Opis ogólny
The coiled condenser circulates coolant through the coil. The jacketed version provides additional cooling capacity by allowing vapors to condense on the inner wall of the jacket.
Informacje prawne
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)