Kluczowe dokumenty
Z529028
VerSA-Flow™ can and drum dispenser
Economy pressure-delivery cap
About This Item
Kod UNSPSC:
41121500
NACRES:
NB.01
Polecane produkty
Powiązane kategorie
Opis ogólny
For low-flow pressure-delivery of solvents and for waste solvent collection using Aldrich cans and drums with tapered 2 inch NPT thread. Use at <5 psi max.
Please refer to Aldrich Technical Information Bulletin AL-210 for additional VerSA-Flow can and drum dispensers and details for using this equipment. This bulletin is available from the Technical Reference section on our website at www.sigma-aldrich.com
- Solid machined PBT body
- Includes 10 ft × 1/4 in. O.D. FEP tubing, adjustable depth through compression fittings on cap
- Can be adapted to 2 in. buttress threads with optional thread adapter Z529036
Please refer to Aldrich Technical Information Bulletin AL-210 for additional VerSA-Flow can and drum dispensers and details for using this equipment. This bulletin is available from the Technical Reference section on our website at www.sigma-aldrich.com
Informacje prawne
VerSA-Flow is a trademark of Sigma-Aldrich Co. LLC
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z529028-1EA | 4061833640746 |