Z509361
Weighing funnel
size XL, anti-static PP
Synonim(y):
Lejek pomiarowy, Lejek skali
About This Item
Kod UNSPSC:
41121809
NACRES:
NB.71
Polecane produkty
Materiały
anti-static PP
rozmiar
XL
opakowanie
pkg of 10 ea
Opis ogólny
- Sit flat on balance - No rolling, no spilling!
- Pouring necks designed to fit volumetric flasks, micro-tubes or any narrow neck apparatus for spill free transfer
- Residual sample can be rinsed to guarantee 100% transfer
- Suitable for safe handling of hazardous materials
- Available in five different sizes: extra small, small, medium, large, and extra large
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)