Kluczowe dokumenty
Z266086
Sigma-Aldrich
TLC desiccating cabinet
provides complete visability of stored TLC plates under desiccating conditions
About This Item
Kod UNSPSC:
41104510
NACRES:
NB.43
Opis ogólny
Acrylic cabinet provides complete visability of stored TLC plates under desiccating conditions. Stores up to 24 (20 × 20 cm) plates. Plates should be oven dried before storage for optimal results.
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)