Kluczowe dokumenty
Z261416
Sigma-Aldrich
Manual gas control valve
stainless steel, CGA Inlet size 330, female ¼ in. outlet
About This Item
Kod UNSPSC:
41112404
NACRES:
NB.51
Materiały
stainless steel
zakres pomiaru ciśnienia wlotowego
≤3,000 psig
rozmiar wlotu kwasu chlorogenowego
330
Opis ogólny
Provides an economical means of controlling gas flow directly from the cylinder. Manufactured with SS or Monel® bodies to accommodate corrosive gases that cannot be run through a regulator. Valve stem packing is PTFE. Hose barb outlet.
Ostrzeżenie
Control valves do not regulate pressure, they only control flow.
Informacje prawne
Monel is a registered trademark of Huntington Alloys Corp.
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)