Kluczowe dokumenty
Z147303
Sigma-Aldrich
Lecture bottle adapter, CGA to NPT
CGA 180 male inlet and outlet for 1/4 in. NPT female, brass
About This Item
Kod UNSPSC:
41100000
NACRES:
NB.51
Materiały
brass
dopasowywanie
CGA 180 male inlet and outlet for 1/4 in. NPT female
Inne uwagi
The adapter connects a lecture-bottle regulator with a CGA 180 female inlet to a needle valve with 1/4 inch NPT male outlet thread.
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)
Odwiedź Bibliotekę dokumentów
Global Trade Item Number
SKUGTIN
Z147303-1EA4061837603389