Z243264
ACE Squibb separatory funnel with Locktight stopper
capacity 125 mL, stopper joint: ST/NS 24/40
About This Item
Kod UNSPSC:
41121809
NACRES:
NB.43
Polecane produkty
Połączenie
joint: ST/NS stopper24/40
producent / nazwa handlowa
Ace Glass 723157
pojemność
125 mL
Powiązane kategorie
Opis ogólny
European style funnel with new leaktight locking stopper that will not fall out during shaking or transportation. The stopper is secured into a screw-threaded top joint via cap and Viton® O-ring for a positive, leaktight seal. Supplied complete with 1:5 PTFE plug, stopper, cap, and O-ring.
Informacje prawne
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z243264-1EA | 4061832362939 |