Z222321
Keck® Joint Clip
standard taper, violet, joint size 12
About This Item
Kod UNSPSC:
41122413
NACRES:
NB.35
Polecane produkty
kolor
violet
Powiązane kategorie
Opis ogólny
Color coded, Delrin® plastic joint clips will not scratch glassware or corrode. Withstand temperatures up to 302 °F (150 °C). Resistant to concentrated alkalis and diluted acids; not resistant to halogen gases or concentrated acids.
Informacje prawne
Delrin is a registered trademark of E. I. du Pont de Nemours and Company
Keck is a registered trademark of Adcuram Lab Technologies GmbH
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z222321-10EA | |
Z222321-12EA | 4061833308073 |