Z197122
3M™ disposable respirator
organic vapor/acid gas, size M
About This Item
Kod UNSPSC:
46182002
Polecane produkty
rozmiar
M
opakowanie
pkg of 1 ea
producent / nazwa handlowa
3M 5000
Opis ogólny
Pre-assembled disposable respirators. Low-profile design provides improved comfort and less vision interference. Approved for: Organic vapor/acid gas and dusts/mists having a TWA not less than 0.05 mg/m3 or 2 mppcf.
NIOSH approved.
Protection factor: Up to 10 × PEL.
NIOSH approved.
Protection factor: Up to 10 × PEL.
Informacje prawne
3M is a trademark of 3M Company
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?