Z186821
All-purpose sampling containers
capacity 4 1/2 oz
About This Item
Kod UNSPSC:
41121800
NACRES:
NB.51
Polecane produkty
Właściwości
lid: no
pojemność
4 1/2 oz
Opis ogólny
Durable and translucent PP containers resist acids, bases, most organic solvents, and common laboratory reagents. Internally molded dual graduations in increments of 1 oz and 20 mL. Economical enough to discard after one use, or strong enough to reuse. Tight-fitting PE lid not included. Nonsterile.
Inne uwagi
Supplied without lid. No lid available.
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)
Klienci oglądali również te produkty
Global Trade Item Number
SKU | GTIN |
---|---|
Z186821-500EA | 4061832331263 |