Z158038
Spilfyter® hazmat spill control, polypropylene
Pillow, capacity 250 mL
About This Item
Kod UNSPSC:
47131905
NACRES:
NB.35
Polecane produkty
opakowanie
pkg of 20 ea
szer. × dł.
10 in. × 10 in.
pojemność
250 mL
Opis ogólny
Spilfyter® products are made from a specially formulated, lint-free, 100% PP fabric that is virtually inert and will absorb any liquid with which they come in contact. They are able to collect up to 10 times their own weight in waste material (depending upon its viscosity) in 30s. Saturated Spilfyter® products can be landfilled, neutralized, or incinerated in accordance with federal, state and local regulations.
Informacje prawne
Spilfyter is a registered trademark of National Packaging Services Corp.
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z158038-20EA | 4061833568200 |