Z150754
Water aspirator pump
Chapman swirling
About This Item
Kod UNSPSC:
41105100
NACRES:
NB.79
Polecane produkty
Długość
137 mm
Powiązane kategorie
Opis ogólny
For greater air-entrainment capacity.
Operates efficiently on both high and low water pressure. Nickel-plated 3/8 in. NPTM water inlet. Ball-type, positive-acting valve in suction tube prevents backflow should pressure be reduced. Max. vacuum is 100 mm Hg (abs).
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z150754-1EA | 4061832329963 |